Product Details
- SNP ID
-
rs78658157
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:231338093 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGAGTAAAGGCTGAAACTAGCTT[C/G]CTGAAAGCTTCGTAGGGCCCGAGCC
- Phenotype
-
MIM: 615283
MIM: 616086
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EXOC8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112263680] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EXOC8
- Gene Name
- exocyst complex component 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175876.4 |
158 |
Intron |
|
|
NP_787072.2 |
- Gene
- SPRTN
- Gene Name
- SprT-like N-terminal domain
View Full Product Details