Product Details
- SNP ID
-
rs552684984
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:42658631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAACGTCCACGAGGTGCTCAGCGC[C/T]GCCTCATTGCTGCAGATGGCTGACA
- Phenotype
-
MIM: 161565
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928323
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147494762] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928323
- Gene Name
- uncharacterized LOC101928323
There are no transcripts associated with this gene.
- Gene
- NKTR
- Gene Name
- natural killer cell triggering receptor
There are no transcripts associated with this gene.
- Gene
- ZBTB47
- Gene Name
- zinc finger and BTB domain containing 47
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145166.3 |
557 |
Silent Mutation |
GCC,GCT |
A92A |
NP_660149.2 |
View Full Product Details