Product Details

SNP ID
rs975657026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176147208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGGGAGAGATAGGAAGCAGGG[A/G]TGAGGGGCAGATGGAGCGGCTACCC
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
There are no transcripts associated with this gene.

Gene
HOXD4
Gene Name
homeobox D4
There are no transcripts associated with this gene.

Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details