Product Details

SNP ID
hCV27878569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66513970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAAGTGTATGTATAATGTTGCT[C/T]GATACCTGTTTTAACAGGCTGATGT
Phenotype
MIM: 180479 MIM: 609984
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL4 PubMed Links

Gene Details

Gene
RPL4
Gene Name
ribosomal protein L4
There are no transcripts associated with this gene.

Gene
ZWILCH
Gene Name
zwilch kinetochore protein
There are no transcripts associated with this gene.

View Full Product Details