Product Details

SNP ID
rs4666997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:180990566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTCATACGTGTGTGTTATTTTTT[A/G]TGTGAAACCACAGGAATGACTACCT
Phenotype
MIM: 604151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2E3 PubMed Links

Gene Details

Gene
UBE2E3
Gene Name
ubiquitin conjugating enzyme E2 E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278554.1 1605 Intron NP_001265483.1
NM_001278555.1 1605 Intron NP_001265484.1
NM_006357.3 1605 Intron NP_006348.1
NM_182678.2 1605 Intron NP_872619.1
XM_005246244.2 1605 Intron XP_005246301.1
XM_011510492.2 1605 UTR 3 XP_011508794.1
XM_017003166.1 1605 Intron XP_016858655.1
XM_017003167.1 1605 Intron XP_016858656.1

View Full Product Details