Product Details
- SNP ID
-
rs4461146
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:12100529 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTGACATGGTGATTTGTGTCACT[C/T]AAAAAGTAACTGTCTTTAAAATAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD62
PubMed Links
Gene Details
- Gene
- ANKRD62
- Gene Name
- ankyrin repeat domain 62
View Full Product Details