Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284395.1 | Intron | NP_001271324.1 | |||
NM_001284396.1 | Intron | NP_001271325.1 | |||
NM_020212.1 | Intron | NP_064597.1 | |||
XM_006720614.1 | Intron | XP_006720677.1 | |||
XM_006720615.1 | Intron | XP_006720678.1 | |||
XM_006720618.2 | Intron | XP_006720681.1 | |||
XM_011521794.2 | Intron | XP_011520096.1 | |||
XM_011521795.1 | Intron | XP_011520097.1 | |||
XM_011521796.2 | Intron | XP_011520098.1 | |||
XM_011521798.2 | Intron | XP_011520100.1 | |||
XM_011521800.1 | Intron | XP_011520102.1 | |||
XM_011521802.2 | Intron | XP_011520104.1 | |||
XM_017022415.1 | Intron | XP_016877904.1 | |||
XM_017022416.1 | Intron | XP_016877905.1 | |||
XM_017022417.1 | Intron | XP_016877906.1 |