Product Details

SNP ID
rs4601793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126213465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGACAGAGGTCTATAAAAGTTGA[A/G]AAATTCCACCCTAAAAGAATTATTA
Phenotype
MIM: 616587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024556.3 Intron NP_078832.1
Gene
RPUSD4
Gene Name
RNA pseudouridylate synthase domain containing 4
There are no transcripts associated with this gene.

View Full Product Details