Product Details

SNP ID
rs4726048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151558703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACACTGTATCATAAGGAATTCCAA[C/T]CCAGCTACCCCCATCTTCACGGCAC
Phenotype
MIM: 602743
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKAG2 PubMed Links

Gene Details

Gene
PRKAG2
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040633.1 3513 Intron NP_001035723.1
NM_001304527.1 3513 Intron NP_001291456.1
NM_001304531.1 3513 Intron NP_001291460.1
NM_016203.3 3513 Intron NP_057287.2
NM_024429.1 3513 Intron NP_077747.1
XM_005250002.3 3513 UTR 3 XP_005250059.1
XM_005250004.3 3513 UTR 3 XP_005250061.1
XM_005250006.4 3513 UTR 3 XP_005250063.1
XM_006716021.2 3513 UTR 3 XP_006716084.1
XM_011516282.1 3513 UTR 3 XP_011514584.1
XM_011516283.1 3513 Intron XP_011514585.1
XM_011516284.1 3513 Intron XP_011514586.1
XM_011516285.1 3513 UTR 3 XP_011514587.1
XM_011516286.1 3513 UTR 3 XP_011514588.1
XM_011516287.1 3513 UTR 3 XP_011514589.1
XM_017012268.1 3513 UTR 3 XP_016867757.1
XM_017012269.1 3513 Intron XP_016867758.1
XM_017012270.1 3513 Intron XP_016867759.1
XM_017012271.1 3513 Intron XP_016867760.1
XM_017012272.1 3513 Intron XP_016867761.1
XM_017012273.1 3513 Intron XP_016867762.1
XM_017012274.1 3513 UTR 3 XP_016867763.1
XM_017012275.1 3513 UTR 3 XP_016867764.1
XM_017012276.1 3513 Intron XP_016867765.1
XM_017012277.1 3513 Intron XP_016867766.1
XM_017012278.1 3513 Intron XP_016867767.1
XM_017012279.1 3513 Intron XP_016867768.1
XM_017012280.1 3513 Intron XP_016867769.1
XM_017012281.1 3513 Intron XP_016867770.1
XM_017012282.1 3513 Intron XP_016867771.1

View Full Product Details