Product Details

SNP ID
rs4820345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38695146 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAATCAGGCATGGGTCAAAGAGC[C/T]AGGCACACATTAGTAATCCGTAGGT
Phenotype
MIM: 602245 MIM: 615323
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTPBP1 PubMed Links

Gene Details

Gene
GTPBP1
Gene Name
GTP binding protein 1
There are no transcripts associated with this gene.

Gene
JOSD1
Gene Name
Josephin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014876.5 Intron NP_055691.1
XM_005261876.2 Intron XP_005261933.1
XM_005261877.2 Intron XP_005261934.1
XM_005261878.2 Intron XP_005261935.1
XM_005261879.2 Intron XP_005261936.1

View Full Product Details