Product Details

SNP ID
rs4844295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71543170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTAAGTGATTTTGGAAACCATAA[C/T]ATAGTTCCAGAGCAAATCATATATA
Phenotype
MIM: 300255
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OGT PubMed Links

Gene Details

Gene
OGT
Gene Name
O-linked N-acetylglucosamine (GlcNAc) transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181672.2 Intron NP_858058.1
NM_181673.2 Intron NP_858059.1

View Full Product Details