Product Details

SNP ID
rs4645511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:137385487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGAGTGGTCAGGTATCCTCTG[A/T]TTAAATGCTAACAGAGGCTGAAAGT
Phenotype
MIM: 604072
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DGKI PubMed Links

Gene Details

Gene
DGKI
Gene Name
diacylglycerol kinase iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321708.1 Intron NP_001308637.1
NM_001321709.1 Intron NP_001308638.1
NM_001321710.1 Intron NP_001308639.1
NM_004717.3 Intron NP_004708.1

View Full Product Details