Product Details

SNP ID
rs7824350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:72540253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATTAGTTCCTTTAAAACTTGCT[A/G]TCTCTTAAAGACTAATTCCAAAGGA
Phenotype
MIM: 607738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNB2 PubMed Links

Gene Details

Gene
KCNB2
Gene Name
potassium voltage-gated channel subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004770.2 Intron NP_004761.2
XM_017013981.1 Intron XP_016869470.1
XM_017013982.1 Intron XP_016869471.1

View Full Product Details