Product Details

SNP ID
rs7213926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75181697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATCTCCATTAAAGGAAACACAC[A/G]AACAAAAAAAACAATCGAAGAATAT
Phenotype
MIM: 603042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUMO2 PubMed Links

Gene Details

Gene
SUMO2
Gene Name
small ubiquitin-like modifier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005849.1 Intron NP_001005849.1
NM_006937.3 Intron NP_008868.3

View Full Product Details