Product Details

SNP ID
rs7041904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128421368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGTAGGTTGGGCCCAGCCAGCCC[C/T]TTTCACCCCAGCCTTCAGGGCCCAG
Phenotype
MIM: 616626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CERCAM PubMed Links

Gene Details

Gene
CERCAM
Gene Name
cerebral endothelial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286760.1 Intron NP_001273689.1
NM_016174.4 Intron NP_057258.3
XM_005252035.1 Intron XP_005252092.1
XM_011518762.1 Intron XP_011517064.1
XM_011518763.2 Intron XP_011517065.1
XM_017014794.1 Intron XP_016870283.1

View Full Product Details