Product Details

SNP ID
rs7927826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65947700 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAGGGAGAAGTGCAGGGCCAGA[A/G]CCAGGGGAGCAGCCCCAGCTTCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSGA10IP PubMed Links

Gene Details

Gene
TSGA10IP
Gene Name
testis specific 10 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152762.2 1106 Missense Mutation AAC,AGC N292S NP_689975.2
XM_011544887.2 1106 Missense Mutation AAC,AGC N292S XP_011543189.1
XM_011544888.2 1106 Missense Mutation AAC,AGC N156S XP_011543190.1

View Full Product Details