Product Details

SNP ID
rs7927956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65947838 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGACCTGGACTGTGGTGAGCGTG[C/G]GGCTCAGAGCCTGGATTCCCCCGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSGA10IP PubMed Links

Gene Details

Gene
TSGA10IP
Gene Name
testis specific 10 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152762.2 Intron NP_689975.2
XM_011544887.2 Intron XP_011543189.1
XM_011544888.2 Intron XP_011543190.1

View Full Product Details