Product Details

SNP ID
rs7077344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96364958 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGTGAATTTGCTAGTTAGCAGG[A/C]TTTTACAAATTATTTAAATCAGCTG
Phenotype
MIM: 606743
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OPALIN PubMed Links

Gene Details

Gene
OPALIN
Gene Name
oligodendrocytic myelin paranodal and inner loop protein
There are no transcripts associated with this gene.

Gene
TLL2
Gene Name
tolloid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012465.3 6419 UTR 3 NP_036597.1

View Full Product Details