Product Details
- SNP ID
-
rs7924834
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:71933287 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGGATGTGTCCACTCTCAAGTCCT[A/G]TTTATCTGCCAAAATGATGTGAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100133315
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74411661] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100133315
- Gene Name
- transient receptor potential cation channel, subfamily C, member 2-like
There are no transcripts associated with this gene.
- Gene
- RNF121
- Gene Name
- ring finger protein 121
View Full Product Details