Product Details
- SNP ID
-
rs4716647
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:156648167 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAAACCGAAAAGTTAGAAATATGA[C/T]TTTTTATTGATAGGCATGGAAAATA
- Phenotype
-
MIM: 610242
MIM: 610241
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C7orf13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75567524] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C7orf13
- Gene Name
- chromosome 7 open reading frame 13
There are no transcripts associated with this gene.
- Gene
- LOC101927858
- Gene Name
- uncharacterized LOC101927858
There are no transcripts associated with this gene.
- Gene
- RNF32
- Gene Name
- ring finger protein 32
View Full Product Details