Product Details

SNP ID
rs5981078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71150098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGAGCGCTTCTAATGCACACC[C/G]GAGCATGGAATGAGACCCGCTGCTG
Phenotype
MIM: 300188 MIM: 300336
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MED12 PubMed Links

Gene Details

Gene
MED12
Gene Name
mediator complex subunit 12
There are no transcripts associated with this gene.

Gene
NLGN3
Gene Name
neuroligin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166660.1 Intron NP_001160132.1
NM_001321276.1 Intron NP_001308205.1
NM_018977.3 Intron NP_061850.2
NM_181303.1 Intron NP_851820.1

View Full Product Details