Product Details

SNP ID
rs6524991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65525561 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCTAGACCAAATATGGTCCATA[A/G]GCCTTATTACTAGGGGTTCTAAAAA
Phenotype
MIM: 300964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAS1L PubMed Links

Gene Details

Gene
LAS1L
Gene Name
LAS1 like, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170649.1 Intron NP_001164120.1
NM_001170650.1 Intron NP_001164121.1
NM_031206.4 Intron NP_112483.1
XM_005262301.1 Intron XP_005262358.1
XM_005262304.1 Intron XP_005262361.1
XM_005262305.3 Intron XP_005262362.1
XM_005262306.3 Intron XP_005262363.1
XM_011531045.1 Intron XP_011529347.1
XM_011531046.1 Intron XP_011529348.1
XM_017029877.1 Intron XP_016885366.1
XM_017029878.1 Intron XP_016885367.1
XM_017029879.1 Intron XP_016885368.1
XM_017029880.1 Intron XP_016885369.1

View Full Product Details