Product Details

SNP ID
rs8109247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32999779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCACGTAGAGATTTCTCTACCA[C/T]GTCTCCAGCTTCCTTTTTTCTGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHPN2 PubMed Links

Gene Details

Gene
RHPN2
Gene Name
rhophilin Rho GTPase binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033103.4 Intron NP_149094.3

View Full Product Details