Product Details

SNP ID
rs9870815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195726334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTGCTTCCTCCTACAGCCCCTC[A/G]GAAGCCGCCCTCAAGAACTTCACCC
Phenotype
MIM: 610360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC20 PubMed Links

Gene Details

Gene
MUC20
Gene Name
mucin 20, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282506.1 1857 Silent Mutation TCA,TCG S577S NP_001269435.1
NM_001291833.1 1857 Silent Mutation TCA,TCG S403S NP_001278762.1
NM_020790.1 1857 Silent Mutation TCA,TCG S423S NP_065841.1
NM_152673.3 1857 Silent Mutation TCA,TCG S406S NP_689886.3

View Full Product Details