Product Details

SNP ID
rs9785892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:20472878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCACATTTAATCCCATTGCATTT[A/C]GCAGATTCTGATTTCAACTTCTAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTTY10 PubMed Links

Gene Details

Gene
TTTY10
Gene Name
testis-specific transcript, Y-linked 10 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details