Product Details

SNP ID
hCV30251242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42659508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCCACACGGTCCCGGGAGAAC[A/G]CCCGGCGCCGGGGTACCCCTGAACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928323 PubMed Links

Gene Details

Gene
LOC101928323
Gene Name
uncharacterized LOC101928323
There are no transcripts associated with this gene.

Gene
ZBTB47
Gene Name
zinc finger and BTB domain containing 47
There are no transcripts associated with this gene.

View Full Product Details