Product Details

SNP ID
rs10262758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139432448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGACATTTACAGAGTTTAAATT[A/G]TATGCAGGCGAAATACACAATCTTG
Phenotype
MIM: 613056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf55-LUC7L2 PubMed Links

Gene Details

Gene
C7orf55-LUC7L2
Gene Name
C7orf55-LUC7L2 readthrough
There are no transcripts associated with this gene.

Gene
KLRG2
Gene Name
killer cell lectin like receptor G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198508.2 Intron NP_940910.1
XM_005250311.3 Intron XP_005250368.1
XM_011516140.2 Intron XP_011514442.1
XM_011516141.2 Intron XP_011514443.1
Gene
LOC100129148
Gene Name
uncharacterized LOC100129148
There are no transcripts associated with this gene.

Gene
LUC7L2
Gene Name
LUC7 like 2, pre-mRNA splicing factor
There are no transcripts associated with this gene.

View Full Product Details