Product Details

SNP ID
rs9614646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45345498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATCAGCGCGTCGGCTCTGGAATA[C/G]AACTCTTCTTTTAGGGAGATGACTA
Phenotype
MIM: 608685
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM118A PubMed Links

Gene Details

Gene
FAM118A
Gene Name
family with sequence similarity 118 member A
There are no transcripts associated with this gene.

Gene
SMC1B
Gene Name
structural maintenance of chromosomes 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291501.1 3519 Missense Mutation TTC,TTG F1115L NP_001278430.1
NM_148674.4 3519 Missense Mutation TTC,TTG F1189L NP_683515.4
XM_011530144.2 3519 Missense Mutation TTC,TTG F1148L XP_011528446.1
XM_011530145.2 3519 UTR 3 XP_011528447.1

View Full Product Details