Product Details

SNP ID
rs9397004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169792040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACCCACCCCCGCTGAGACCGGC[A/G]GATTCTCTCCCTCCCCACTTGAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00242 PubMed Links

Gene Details

Gene
LINC00242
Gene Name
long intergenic non-protein coding RNA 242
There are no transcripts associated with this gene.

Gene
LINC00574
Gene Name
long intergenic non-protein coding RNA 574
There are no transcripts associated with this gene.

View Full Product Details