Product Details

SNP ID
rs10497094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:152119488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTGACAGCTTAGCAACGGTACTT[C/G]AATCACCTCTTTAAAAAAACAACAC
Phenotype
MIM: 606244
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STAM2 PubMed Links

Gene Details

Gene
STAM2
Gene Name
signal transducing adaptor molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005843.5 3014 UTR 3 NP_005834.4

View Full Product Details