Product Details

SNP ID
rs10009883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165032847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTGAGATCTGGGAATCTCCCAC[A/C]GGTTTGATTTATATTCACTGAGCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRIM60 PubMed Links

Gene Details

Gene
TRIM60
Gene Name
tripartite motif containing 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258025.1 Intron NP_001244954.1
NM_152620.2 Intron NP_689833.1
XM_011531683.2 Intron XP_011529985.1

View Full Product Details