Product Details

SNP ID
rs10412551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46410492 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCCGGGGCATGAGGATGGCAAGC[A/G]AAGTGACAACATAAACACAGGTGTC
Phenotype
MIM: 614145
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC8 PubMed Links

Gene Details

Gene
CCDC8
Gene Name
coiled-coil domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032040.4 3171 UTR 3 NP_114429.2

View Full Product Details