Product Details

SNP ID
rs6057768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33221144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGAAAGTTTTTATGATGTACT[A/G]TTAGATTTTTTTTTTTTTTTTTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFA3 PubMed Links

Gene Details

Gene
BPIFA3
Gene Name
BPI fold containing family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042439.2 Intron NP_001035904.1
NM_178466.4 Intron NP_848561.2

View Full Product Details