Product Details

SNP ID
rs9382101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52363992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTGTATTGTTAATTATGTATG[A/G]TAATTATTTCAGTTATTTGGCGTAA
Phenotype
MIM: 607780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAQR8 PubMed Links

Gene Details

Gene
PAQR8
Gene Name
progestin and adipoQ receptor family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133367.4 Intron NP_588608.1

View Full Product Details