Product Details

SNP ID
rs10241658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2359462 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGACCAGGTTCAGGGGCTCCATG[C/T]AGCGGAACAGGTGGAGGCTTCTGCA
Phenotype
MIM: 603917 MIM: 614905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3B PubMed Links

Gene Details

Gene
EIF3B
Gene Name
eukaryotic translation initiation factor 3 subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037283.1 Intron NP_001032360.1
NM_003751.3 Intron NP_003742.2
XM_011515599.1 Intron XP_011513901.1
XM_011515600.1 Intron XP_011513902.1
XM_011515601.1 Intron XP_011513903.1
XM_017012752.1 Intron XP_016868241.1
XM_017012753.1 Intron XP_016868242.1
Gene
SNX8
Gene Name
sorting nexin 8
There are no transcripts associated with this gene.

View Full Product Details