Product Details

SNP ID
rs13306752
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:42927419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTAGAATGTAACAGGCTCCAG[A/G]AGCACCATTCAAGGCTAAGGGGAGA
Phenotype
MIM: 138140 MIM: 143090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC2A1 PubMed Links

Gene Details

Gene
SLC2A1
Gene Name
solute carrier family 2 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006516.2 Intron NP_006507.2

View Full Product Details