Product Details
- SNP ID
-
rs12640030
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:151672320 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTTTTCCCTTCTGCCTTTCTTCC[C/G]ACCTCTTAGAAGCATGAAGCTGAAG
- Phenotype
-
MIM: 603645
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM160A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79150013] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM160A1
- Gene Name
- family with sequence similarity 160 member A1
There are no transcripts associated with this gene.
- Gene
- GATB
- Gene Name
- glutamyl-tRNA amidotransferase subunit B
View Full Product Details