Product Details

SNP ID
rs13129926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151670601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCAGTCGATAGGTTGATTAATT[C/T]TAATAATATCCTCCTTCCCCACCCT
Phenotype
MIM: 603645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM160A1 PubMed Links

Gene Details

Gene
FAM160A1
Gene Name
family with sequence similarity 160 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109977.1 2115 Intron NP_001103447.1
XM_005263199.1 2115 Intron XP_005263256.1
XM_011532220.1 2115 Intron XP_011530522.1
XM_011532221.2 2115 Intron XP_011530523.1
XM_011532222.2 2115 Intron XP_011530524.1
XM_011532223.2 2115 Intron XP_011530525.1
XM_011532224.2 2115 Intron XP_011530526.1
XM_017008583.1 2115 Intron XP_016864072.1
Gene
GATB
Gene Name
glutamyl-tRNA amidotransferase subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004564.2 2115 Intron NP_004555.1
XM_005263063.3 2115 UTR 3 XP_005263120.1

View Full Product Details