Product Details
- SNP ID
-
rs11750712
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:31533289 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAAAAAAAAATTTAACACGG[G/T]GCCCTGCATAGATAAACACTAAAAG
- Phenotype
-
MIM: 608828
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C5orf22
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5867085] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C5orf22
- Gene Name
- chromosome 5 open reading frame 22
- Gene
- DROSHA
- Gene Name
- drosha ribonuclease III
There are no transcripts associated with this gene.
View Full Product Details