Product Details

SNP ID
rs7112925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67058689 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAGCATCTGAGCACAAGCAGGA[C/T]GGTCGGGCCTACCCCAGTTCTTATC
Phenotype
MIM: 605781 MIM: 606436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHOD PubMed Links

Gene Details

Gene
RHOD
Gene Name
ras homolog family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300886.1 Intron NP_001287815.1
NM_014578.3 Intron NP_055393.1
Gene
SYT12
Gene Name
synaptotagmin 12
There are no transcripts associated with this gene.

View Full Product Details