Product Details

SNP ID
rs12611091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55288961 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCACAGAAGCTGGCTTGGACTGG[C/T]GCAACCTTGCAATCGTTAATACTCG
Phenotype
MIM: 609235 MIM: 612939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRSK1 PubMed Links

Gene Details

Gene
BRSK1
Gene Name
BR serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032430.1 Intron NP_115806.1
XM_005259327.3 Intron XP_005259384.1
XM_011527395.2 Intron XP_011525697.2
Gene
HSPBP1
Gene Name
HSPA (Hsp70) binding protein 1
There are no transcripts associated with this gene.

View Full Product Details