Product Details

SNP ID
rs4889565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31885987 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGAGCTTAGAACATTGTCCAGT[G/A]TATAATAAACTCTAAACTGTTGCCT
Phenotype
MIM: 604752
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNF267 PubMed Links

Gene Details

Gene
ZNF267
Gene Name
zinc finger protein 267
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265588.1 Intron NP_001252517.1
NM_003414.5 Intron NP_003405.3
XM_017022865.1 Intron XP_016878354.1
XM_017022866.1 Intron XP_016878355.1

View Full Product Details