Product Details

SNP ID
rs6873370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140646489 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGTGTCTCACAGATATCTACTCT[C/G]AGAACCAATCTGCTTAAGGTGGTCA
Phenotype
MIM: 600549 MIM: 602137
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IK PubMed Links

Gene Details

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
There are no transcripts associated with this gene.

Gene
MIR3655
Gene Name
microRNA 3655
There are no transcripts associated with this gene.

Gene
NDUFA2
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185012.1 Intron NP_001171941.1
NM_002488.4 Intron NP_002479.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 Intron NP_001287909.1
NM_001300982.1 Intron NP_001287911.1
NM_018502.4 Intron NP_060972.3
XM_005268477.1 Intron XP_005268534.1
XM_011537663.2 Intron XP_011535965.1
XM_011537665.2 Intron XP_011535967.1
XM_011537668.2 Intron XP_011535970.1
XM_017009617.1 Intron XP_016865106.1
XM_017009618.1 Intron XP_016865107.1
XM_017009619.1 Intron XP_016865108.1
XM_017009620.1 Intron XP_016865109.1

View Full Product Details