Product Details

SNP ID
rs12933355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53713838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGCAGATCTCTGGCACTGATTC[A/T]TCTTGTATAATTGAAACTTTATGCC
Phenotype
MIM: 610966 MIM: 610937
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FTO PubMed Links

Gene Details

Gene
FTO
Gene Name
fat mass and obesity associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080432.2 Intron NP_001073901.1
XM_011523313.2 Intron XP_011521615.1
XM_011523314.2 Intron XP_011521616.1
XM_011523315.2 Intron XP_011521617.1
XM_011523316.2 Intron XP_011521618.1
XM_017023654.1 Intron XP_016879143.1
XM_017023655.1 Intron XP_016879144.1
XM_017023656.1 Intron XP_016879145.1
XM_017023657.1 Intron XP_016879146.1
XM_017023658.1 Intron XP_016879147.1
Gene
RPGRIP1L
Gene Name
RPGRIP1 like
There are no transcripts associated with this gene.

View Full Product Details