Product Details

SNP ID
rs6925514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110832745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCTCACTCTTAGATTACCCCTC[C/T]TTGTACTTCTGAATCTATTTCAGTG
Phenotype
MIM: 180980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMD1 PubMed Links

Gene Details

Gene
AMD1
Gene Name
adenosylmethionine decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287214.1 Intron NP_001274143.1
NM_001287215.1 Intron NP_001274144.1
NM_001287216.1 Intron NP_001274145.1
NM_001634.5 Intron NP_001625.2

View Full Product Details