Product Details

SNP ID
rs12118668
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:229519415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCGTTCATAGCACAGTTTTATC[C/T]TGCCATCAGAGTAATCACTAGACCG
Phenotype
MIM: 605454
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB10 PubMed Links

Gene Details

Gene
ABCB10
Gene Name
ATP binding cassette subfamily B member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012089.2 Intron NP_036221.2
XM_011544135.2 Intron XP_011542437.1
XM_011544136.2 Intron XP_011542438.1
Gene
LOC101927478
Gene Name
uncharacterized LOC101927478
There are no transcripts associated with this gene.

View Full Product Details