Product Details

SNP ID
rs8023458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43788741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGGTTAGACTCTCAGCTCCACC[C/T]CTTATTATCTCAGTTTCGTTGAGAA
Phenotype
MIM: 605054 MIM: 614550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1282 PubMed Links

Gene Details

Gene
MIR1282
Gene Name
microRNA 1282
There are no transcripts associated with this gene.

Gene
SERF2
Gene Name
small EDRK-rich factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018108.3 Intron NP_001018118.1
NM_001199875.1 Intron NP_001186804.1
NM_001199876.1 Intron NP_001186805.1
NM_001199877.1 Intron NP_001186806.1
NM_001199878.1 Intron NP_001186807.1
Gene
SERF2-C15ORF63
Gene Name
SERF2-C15orf63 readthrough
There are no transcripts associated with this gene.

Gene
SERINC4
Gene Name
serine incorporator 4
There are no transcripts associated with this gene.

View Full Product Details