Product Details

SNP ID
rs6778518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98576589 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAAGCCATTTACAAATGTCTAAG[C/T]GTCACTAACTTAAATGTGCTAATTT
Phenotype
MIM: 612732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPOX PubMed Links

Gene Details

Gene
CPOX
Gene Name
coproporphyrinogen oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000097.5 3025 Intron NP_000088.3
XM_005247125.4 3025 UTR 3 XP_005247182.1

View Full Product Details