Product Details

SNP ID
hCV31533112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:52776061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCATGCACCTATAGTCCCAGCC[A/T]CATGGGAGGCTGAGGTGGAAGGATC
Phenotype
MIM: 616510 MIM: 615814
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GNPNAT1 PubMed Links

Gene Details

Gene
GNPNAT1
Gene Name
glucosamine-phosphate N-acetyltransferase 1
There are no transcripts associated with this gene.

Gene
STYX
Gene Name
serine/threonine/tyrosine interacting protein
There are no transcripts associated with this gene.

View Full Product Details