Product Details

SNP ID
rs11654663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67393189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATACACATAACATTTGCCATCTT[A/C]AAACTTTTCGAGTGTACAGTTTCAG
Phenotype
MIM: 605134
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PITPNC1 PubMed Links

Gene Details

Gene
PITPNC1
Gene Name
phosphatidylinositol transfer protein, cytoplasmic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012417.3 Intron NP_036549.2
NM_181671.2 Intron NP_858057.1
XM_005257216.2 Intron XP_005257273.1
XM_017024442.1 Intron XP_016879931.1
XM_017024443.1 Intron XP_016879932.1
XM_017024444.1 Intron XP_016879933.1
XM_017024445.1 Intron XP_016879934.1

View Full Product Details